Anti-PCNA Rabbit Polyclonal Antibody

Supplier: Abnova
PAB12722
10733-406EA 406.15 USD
10733-406
Anti-PCNA Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against recombinant PCNA.

Recommended Dilutions: Western Blot: 0.1-1 ?g/ml; Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 2-5 ?g/ml

Type: Primary
Antigen: PCNA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR