Anti-CNTN3 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB20194
10711-452EA 428.55 USD
10711-452
Anti-CNTN3 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against recombinant CNTN3.

Recommended Dilutions: Immunohistochemistry: 1:50-1:200

Type: Primary
Antigen: CNTN3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR