Anti-HYPM Rabbit Polyclonal Antibody

Supplier: Abnova
PAB20197
10709-644EA 527.69 USD
10709-644
Anti-HYPM Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against recombinant CXorf27.

Recommended Dilutions: Immunohistochemistry: 1:50-1:200

Type: Primary
Antigen: HYPM
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR