Anti-immunodeficiency virus type 2 gp36 Mouse Monoclonal Antibody [clone: BDI411]

Supplier: Abnova
MAB4817
10717-498EA 849 USD
10717-498
Anti-immunodeficiency virus type 2 gp36 Mouse Monoclonal Antibody [clone: BDI411]
Antibodies
Mouse monoclonal antibody raised against partial recombinant human immunodeficiency virus type 2 gp36.

Recommended Dilutions: Western Blot: 1:1000; ELISA: 1:500

Type: Primary
Antigen: immunodeficiency virus type 2 gp36
Clonality: Monoclonal
Clone: BDI411
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Viruses
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR