Anti-AFB1 Monoclonal Antibody [clone: 6A1]

Supplier: Abnova
MAB2406
10688-368EA 530.32 USD
10688-368
Anti-AFB1 Monoclonal Antibody [clone: 6A1]
Antibodies
Mouse monoclonal antibody raised against Aflatoxin B1.

Recommended Dilutions: Immunohistochemistry: 1:50-1:100

Type: Primary
Antigen: AFB1
Clonality: Monoclonal
Clone: 6A1
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG1 kappa
Reactivity: Fungi
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR