Anti-CORIN Rabbit Polyclonal Antibody

Supplier: Abnova
PAB12762
10708-824EA 329 USD
10708-824
Anti-CORIN Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of Corin.

Recommended Dilutions: Western Blot: 0.1-1 ?g/ml; ELISA: 0.01-0.1 ?g/ml; Immunoprecipitation: 2-5 ?g/ml; Immunohistochemistry: 0.5-2 ?g/ml

Type: Primary
Antigen: CORIN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR