Anti-ABCA1 Rat Monoclonal Antibody [clone: 5A1-1422.11]

Supplier: Abnova
MAB2419
10687-576EA 530.32 USD
10687-576
Anti-ABCA1 Rat Monoclonal Antibody [clone: 5A1-1422.11]
Antibodies
Rat monoclonal antibody raised against recombinant Abca1.

Recommended Dilutions: Flow Cytometry: 1:1000-1:30000

Type: Primary
Antigen: ABCA1
Clonality: Monoclonal
Clone: 5A1-1422.11
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a kappa
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR