Anti-ABCA1 Rat Monoclonal Antibody [clone: 3A1.891.3]

Supplier: Abnova
MAB2420
10687-574EA 530.14 USD
10687-574
Anti-ABCA1 Rat Monoclonal Antibody [clone: 3A1.891.3]
Antibodies
Rat monoclonal antibody raised against synthetic peptide of Abca1.

Recommended Dilutions: Western Blot: 1:1000-1:3000

Type: Primary
Antigen: ABCA1
Clonality: Monoclonal
Clone: 3A1.891.3
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1 kappa
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR