Anti-CAM Mouse Monoclonal Antibody [clone: CM-16]

Supplier: Abnova
MAB2422
10704-776EA 427.45 USD
10704-776
Anti-CAM Mouse Monoclonal Antibody [clone: CM-16]
Antibodies
Mouse monoclonal antibody raised against Calmodulin.

Recommended Dilutions: Western Blot: 0.5 ?g/ml

Type: Primary
Antigen: CAM
Clonality: Monoclonal
Clone: CM-16
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Chicken, Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR