Anti-C1QTNF2 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB13421
10704-140EA 428 USD
10704-140
Anti-C1QTNF2 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of C1QTNF2.

Recommended Dilutions: Western Blot: 1 ?g/ml

Type: Primary
Antigen: C1QTNF2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR