Anti-C1QTNF7 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB13423
10704-978EA 428 USD
10704-978
Anti-C1QTNF7 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of C1QTNF7.

Recommended Dilutions: Western Blot: 0.5 ?g/ml

Type: Primary
Antigen: C1QTNF7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR