Anti-immunodeficiency virus type 1 Vif Mouse Monoclonal Antibody [clone: 319]

Supplier: Abnova
MAB2431
10720-950EA 531 USD
10720-950
Anti-immunodeficiency virus type 1 Vif Mouse Monoclonal Antibody [clone: 319]
Antibodies
Mouse monoclonal antibody raised against recombinant human immunodeficiency virus type 1 Vif.

Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: immunodeficiency virus type 1 Vif
Clonality: Monoclonal
Clone: 319
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Viruses
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR