Anti-Methicillin resistant Staphylococcus aureus Mouse Monoclonal Antibody [clone: AC10]

Supplier: Abnova
MAB2444
10725-904EA 531 USD
10725-904
Anti-Methicillin resistant Staphylococcus aureus Mouse Monoclonal Antibody [clone: AC10]
Antibodies
Mouse monoclonal antibody raised against native Methicillin resistant Staphylococcus aureus.

Type: Primary
Antigen: Methicillin resistant Staphylococcus aureus
Clonality: Unconjugated
Clone: Monoclonal
Conjugation: AC10
Epitope:
Host: IgG2a
Isotype: Mouse
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR