Anti-ACE2 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB13445
10687-830EA 527.69 USD
10687-830
Anti-ACE2 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of ACE2.

Recommended Dilutions: Western Blot: 0.5-2 ?g/ml

Type: Primary
Antigen: ACE2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR