Anti-Serratia marcescens Mouse Monoclonal Antibody [clone: B/N4N]

Supplier: Abnova
MAB2451
10736-916EA 531 USD
10736-916
Anti-Serratia marcescens Mouse Monoclonal Antibody [clone: B/N4N]
Antibodies
Mouse monoclonal antibody raised against native Serratia marcescens.

Type: Primary
Antigen: Serratia marcescens
Clonality: Unconjugated
Clone: Monoclonal
Conjugation: B/N4N
Epitope:
Host: IgG1
Isotype: Mouse
Reactivity: S. marcescens
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR