Anti-ZAP70 Mouse Monoclonal Antibody [clone: SBZAP]

Supplier: Abnova
MAB3324
10745-012EA 329 USD
10745-012
Anti-ZAP70 Mouse Monoclonal Antibody [clone: SBZAP]
Antibodies
Mouse monoclonal antibody raised against partial recombinant ZAP70.

Recommended Dilutions: Flow Cytometry: 1 ?g/10^6cells

Type: Primary
Antigen: ZAP70
Clonality: Monoclonal
Clone: SBZAP
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR