Anti-MMP9 Mouse Monoclonal Antibody [clone: SB15c]

Supplier: Abnova
MAB3332
10725-676EA 329 USD
10725-676
Anti-MMP9 Mouse Monoclonal Antibody [clone: SB15c]
Antibodies
Mouse monoclonal antibody raised against recombinant MMP9.

Recommended Dilutions: ELISA: 1:3000-1:8000; Immunohistochemistry: 2 ?g/ml

Type: Primary
Antigen: MMP9
Clonality: Monoclonal
Clone: SB15c
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR