Anti-GFAP Mouse Monoclonal Antibody [clone: SB61b]

Supplier: Abnova
MAB3336
10735-326EA 329 USD
10735-326
Anti-GFAP Mouse Monoclonal Antibody [clone: SB61b]
Antibodies
Mouse monoclonal antibody raised against recombinant GFAP.

Recommended Dilutions: Western Blot: 2 ?g/ml; Immunohistochemistry: 5 ?g/ml

Type: Primary
Antigen: GFAP
Clonality: Unconjugated
Clone: Monoclonal
Conjugation: SB61b
Epitope:
Host: IgG2b
Isotype: Mouse
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR