Anti-NOP1 Mouse Monoclonal Antibody [clone: 28F2]

Supplier: Abnova
MAB3338
10676-920EA 638 USD
10676-920
Anti-NOP1 Mouse Monoclonal Antibody [clone: 28F2]
Antibodies
Mouse monoclonal antibody raised against native NOP1.

Recommended Dilutions: Western Blot: 1:500-1:5000; Immunocytochemistry: 1:1000-1:10000; Immunofluorescence: 1:1000-1:10000

Type: Primary
Antigen: NOP1
Clonality: Unconjugated
Clone: Monoclonal
Conjugation: 28F2
Epitope:
Host: IgG3
Isotype: Mouse
Reactivity: Yeast
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR