Anti-PRPH Mouse Monoclonal Antibody [clone: 7C5]

Supplier: Abnova
MAB3348
10677-118EA 638 USD
10677-118
Anti-PRPH Mouse Monoclonal Antibody [clone: 7C5]
Antibodies
Mouse monoclonal antibody raised against recombinant Prph.

Recommended Dilutions: Immunocytochemistry: 1:500-1:5000; Immunofluorescence: 1:500-1:5000; Western Blot: 1:500-1:2500

Type: Primary
Antigen: PRPH
Clonality: Monoclonal
Clone: 7C5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Chicken, Mammals, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR