Anti-PMA1 Mouse Monoclonal Antibody [clone: 40B7]

Supplier: Abnova
MAB3349
10677-056EA 638 USD
10677-056
Anti-PMA1 Mouse Monoclonal Antibody [clone: 40B7]
Antibodies
Mouse monoclonal antibody raised against native PMA1.

Recommended Dilutions: Western Blot: 1:500-1:5000; Immunocytochemistry: 1:250-1:2500; Immunofluorescence: 1:250-1:2500

Type: Primary
Antigen: PMA1
Clonality: Monoclonal
Clone: 40B7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Yeast
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR