Anti-FSHB Mouse Monoclonal Antibody [clone: 090-10243]

Supplier: Abnova
MAB4074
10627-462EA 746.96 USD
10627-462
Anti-FSHB Mouse Monoclonal Antibody [clone: 090-10243]
Antibodies
Mouse monoclonal antibody raised against FSHB.

Type: Primary
Antigen: FSHB
Clonality: Unconjugated
Clone: Monoclonal
Conjugation: 090-10243
Epitope:
Host: IgG1 kappa
Isotype: Mouse
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR