Anti-GPX3 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB12422
10716-334EA 530.32 USD
10716-334
Anti-GPX3 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against full length recombinant GPX3.

Recommended Dilutions: Immunocytochemistry: 1:200; Immunofluorescence: 1:200; Western Blot: 0.5 ?g/ml

Type: Primary
Antigen: GPX3
Clonality: Unconjugated
Clone: Polyclonal
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human, Primates, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR