Anti-CBX alpha Mouse Monoclonal Antibody [clone: B365M]

Supplier: Abnova
MAB4095
10626-736EA 957.5 USD
10626-736
Anti-CBX alpha Mouse Monoclonal Antibody [clone: B365M]
Antibodies
Mouse monoclonal antibody raised against synthetic peptide of Clostridium botulinum Toxin A.

Recommended Dilutions: Immunocytochemistry: 1:50

Type: Primary
Antigen: CBX alpha
Clonality: Monoclonal
Clone: B365M
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR