Anti-DLX4 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB12448
10710-912EA 653.85 USD
10710-912
Anti-DLX4 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of DLX4.

Recommended Dilutions: Immunohistochemistry: 1:50-1:100

Type: Primary
Antigen: DLX4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR