Anti-PITPNM2 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB20209
10733-580EA 428.55 USD
10733-580
Anti-PITPNM2 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against recombinant PITPNM2.

Recommended Dilutions: Immunohistochemistry: 1:50-1:200; Immunofluorescence: 1-4 ?g/ml

Type: Primary
Antigen: PITPNM2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR