Anti-FAH Rabbit Polyclonal Antibody

Supplier: Abnova
PAB12766
10712-972EA 329 USD
10712-972
Anti-FAH Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against full length recombinant Fah.

Recommended Dilutions: Western Blot: 0.1-1 ?g/ml; ELISA: 0.01-0.1 ?g/ml; Immunoprecipitation: 2-5 ?g/ml; Immunohistochemistry: 2-5 ?g/ml

Type: Primary
Antigen: Fah
Clonality: Unconjugated
Clone: Polyclonal
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR