Anti-Nid1 Rat Monoclonal Antibody [clone: JF2]

Supplier: Abnova
MAB2454
10727-172EA 531 USD
10727-172
Anti-Nid1 Rat Monoclonal Antibody [clone: JF2]
Antibodies
Rat monoclonal antibody raised against partial recombinant Nid1.

Type: Primary
Antigen: Nid1
Clonality: Unconjugated
Clone: Monoclonal
Conjugation: JF2
Epitope:
Host: IgG2a
Isotype: Rat
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR