Anti-LRRC37B Rabbit Polyclonal Antibody

Supplier: Abnova
PAB20799
10724-036EA 428.55 USD
10724-036
Anti-LRRC37B Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against recombinant LRRC37B.

Recommended Dilutions: Immunohistochemistry: 1:50-1:200; Immunofluorescence: 1-4 ?g/ml

Type: Primary
Antigen: LRRC37B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR