Anti-ALYREF Mouse Monoclonal Antibody [clone: 11G5]

Supplier: Abnova
MAB2468
10688-902EA 531 USD
10688-902
Anti-ALYREF Mouse Monoclonal Antibody [clone: 11G5]
Antibodies
Mouse monoclonal antibody raised against recombinant ALYREF.

Type: Primary
Antigen: ALYREF
Clonality: Monoclonal
Clone: 11G5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Clawed frog, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR