Anti-TXNDC15 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB20811
10743-432EA 428.55 USD
10743-432
Anti-TXNDC15 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against recombinant TXNDC15.

Recommended Dilutions: Immunohistochemistry: 1:10-1:20

Type: Primary
Antigen: TXNDC15
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR