Anti-NR1H3 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB13685
10733-118EA 427.45 USD
10733-118
Anti-NR1H3 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of NR1H3.

Recommended Dilutions: Western Blot: 1 ?g/ml

Type: Primary
Antigen: NR1H3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR