Anti-S100 B Rabbit Polyclonal Antibody

Supplier: Abnova
PAB13687
10732-206EA 427.45 USD
10732-206
Anti-S100 B Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of S100B.

Recommended Dilutions: Western Blot: 1 ?g/ml; Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 1 ?g/ml

Type: Primary
Antigen: S100 B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR