Anti-TFPI2 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB13690
10740-990EA 428.44 USD
10740-990
Anti-TFPI2 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of TFPI2.

Recommended Dilutions: Western Blot: 1 ?g/ml; Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 2 ?g/ml

Type: Primary
Antigen: TFPI2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR