Anti-HNRNPA2B1 Mouse Monoclonal Antibody [clone: DP3B3]

Supplier: Abnova
MAB2497
10717-684EA 531 USD
10717-684
Anti-HNRNPA2B1 Mouse Monoclonal Antibody [clone: DP3B3]
Antibodies
Mouse monoclonal antibody raised against recombinant HNRNPA2B1.

Recommended Dilutions: Western Blot: 1:500

Type: Primary
Antigen: HNRNPA2B1
Clonality: Monoclonal
Clone: DP3B3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Chicken, Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR