Anti-COL1A1 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB13714
10627-088EA 849.43 USD
10627-088
Anti-COL1A1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against native Col1a1/Col1a2.

Recommended Dilutions: Immunohistochemistry (Frozen sections): 1:10-1:20

Type: Primary
Antigen: COL1A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR