Anti-LMNA Mouse Monoclonal Antibody [clone: JoL5]

Supplier: Abnova
MAB2502
10637-048EA 531 USD
10637-048
Anti-LMNA Mouse Monoclonal Antibody [clone: JoL5]
Antibodies
Mouse monoclonal antibody raised against recombinant LMNA.

Recommended Dilutions: Western Blot: 1:50; Immunofluorescence: 1:10

Type: Primary
Antigen: LMNA
Clonality: Monoclonal
Clone: JoL5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR