Anti-CGB Mouse Monoclonal Antibody [clone: WM16/71]

Supplier: Abnova
MAB2506
10710-060EA 531 USD
10710-060
Anti-CGB Mouse Monoclonal Antibody [clone: WM16/71]
Antibodies
Mouse monoclonal antibody raised against CGB.

Type: Primary
Antigen: CGB
Clonality: Monoclonal
Clone: WM16/71
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR