Anti-Collagen Type I, II, III, IV, V Rabbit Polyclonal Antibody

Supplier: Abnova
PAB13718
10627-106EA 746.62 USD
10627-106
Anti-Collagen Type I, II, III, IV, V Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against full length native Collagen Type I, II, III, IV, V.

Recommended Dilutions: ELISA: 1:200

Type: Primary
Antigen: Collagen Type I, II, III, IV, V
Clonality: Unconjugated
Clone: Polyclonal
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR