Anti-TNFSF13 Rat Monoclonal Antibody [clone: Sacha-2]

Supplier: Abnova
MAB3365
10745-226EA 1061.25 USD
10745-226
Anti-TNFSF13 Rat Monoclonal Antibody [clone: Sacha-2]
Antibodies
Rat monoclonal antibody raised against partial recombinant TNFSF13.

Type: Primary
Antigen: TNFSF13
Clonality: Monoclonal
Clone: Sacha-2
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR