Anti-LAMA1 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB13719
10681-152EA 638 USD
10681-152
Anti-LAMA1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against full length native Lama1.

Recommended Dilutions: Western Blot: 1:1000-1:5000; Immunocytochemistry: 1:100-1:2500; Immunofluorescence: 1:100-1:2500

Type: Primary
Antigen: LAMA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR