Anti-GFAP Rabbit Polyclonal Antibody

Supplier: Abnova
PAB13721
10715-786EA 786.15 USD
10715-786
Anti-GFAP Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against recombinant GFAP.

Recommended Dilutions: Western Blot: 1:1000-1:5000; Immunocytochemistry: 1:1000-1:10000; Immunofluorescence: 1:1000-1:10000

Type: Primary
Antigen: GFAP
Clonality: Unconjugated
Clone: Polyclonal
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Bovine, Mammals
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR