Anti-ANXA5 Mouse Monoclonal Antibody [clone: VAA-33]

Supplier: Abnova
MAB3391
10689-448EA 747 USD
10689-448
Anti-ANXA5 Mouse Monoclonal Antibody [clone: VAA-33]
Antibodies
Mouse monoclonal antibody raised against recombinant ANXA5.

Type: Primary
Antigen: ANXA5
Clonality: Monoclonal
Clone: VAA-33
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR