Anti-CAV2 0 Polyclonal Antibody

Supplier: Abnova
PAB13739
10687-104EA 786.15 USD
10687-104
Anti-CAV2 0 Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic phosphopeptide of CAV2.

Recommended Dilutions: Western Blot: 0.1-1.0 ?g/ml

Type: Primary
Antigen: CAV2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: 0
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR