Anti-PROCR Rat Monoclonal Antibody [clone: LMR-42]

Supplier: Abnova
MAB3398
10628-432EA 1306.15 USD
10628-432
Anti-PROCR Rat Monoclonal Antibody [clone: LMR-42]
Antibodies
Rat monoclonal antibody raised against full length recombinant PROCR.

Recommended Dilutions: Immunohistochemistry (Frozen sections): 1:20

Type: Primary
Antigen: PROCR
Clonality: Monoclonal
Clone: LMR-42
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR