Anti-VPS13A Rabbit Polyclonal Antibody

Supplier: Abnova
PAB21442
10744-268EA 428.55 USD
10744-268
Anti-VPS13A Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against recombinant VPS13A.

Recommended Dilutions: Immunohistochemistry: 1:50-1:200

Type: Primary
Antigen: VPS13A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR