Anti-CD36 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB12464
10706-410EA 530.32 USD
10706-410
Anti-CD36 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of CD36.

Recommended Dilutions: Immunocytochemistry: 1:50-1-200; Immunofluorescence: 1:50-1:200; Western Blot: 1:2000-1:10000

Type: Primary
Antigen: CD36
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR