Anti-C4ORF3 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB21987
10704-346EA 428.55 USD
10704-346
Anti-C4ORF3 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against recombinant C4orf3.

Recommended Dilutions: Immunohistochemistry: 1:500-1:1000

Type: Primary
Antigen: C4ORF3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR