Anti-ATP13A2 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB12479
10690-438EA 530.32 USD
10690-438
Anti-ATP13A2 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of ATP13A2.

Recommended Dilutions: Western Blot: 2 ?g/ml; Immunohistochemistry: 10-20 ?g/ml

Type: Primary
Antigen: ATP13A2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR