Anti-ZFX Rabbit Polyclonal Antibody

Supplier: Abnova
PAB20245
10745-356EA 428.55 USD
10745-356
Anti-ZFX Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against recombinant ZFX.

  • Recommended Dilutions: Immunohistochemistry: 1:200-1:500; Immunofluorescence: 1-4 μg/ml

Type: Primary
Antigen: ZFX
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR